Learn More
SPEX Certiprep 1ML Perfluorooctanesulfonic acid, 1,000 µg/mL Organic Standard
Brand: SPEX Certiprep S-5059
1ML Perfluorooctanesulfonic acid, 1,000 µg/mL Organic Standard
Safety and Handling
- Perfluorooctanesulfonic acid
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Carcinogenicity Category 2
- Flammable liquid Category 2
- Reproductive toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H351-Suspected of causing cancer.
- H360-May damage fertility or the unborn child.
- H362-May cause harm to breast-fed children.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : methanol, perfluorooctane sulfonic acid