missing translation for 'onlineSavingsMsg'
Learn More

Perazine Sulphoxide 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30458076
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30458076

Brand: LoGiCal LGCAMP0157.0101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Perazine Sulphoxide 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Perazine Sulphoxide
CAS 20627-44-5
Analyte or Component Names Perazine Sulphoxide
Shipping Condition Ice Pack (-20°C)
Molecular Formula C20 H25 N3 O S
Certifications/Compliance ISO 17034
Synonym Perazine Sulphoxide
InChI Formula InChI=1 S/C20H25N3OS/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)25(24)20-10-5-3-8-18(20)23/h2-5,7-10 H,6,11-16H2,1H3
SMILES CN1CCN(CCCN2c3ccccc3S(=O)c4ccccc24)CC1
IUPAC Name 10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine 5-oxide
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 355.5
Quantity 1 mL
Formula Weight 355.1718 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.