missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences Strep-Tactin™ Sepharose™ Column
Shop All IBA Lifesciences Products
Click to view available options
Capacity (Metric):
0.2 mL
1 mL
Unit Size:
1mL
Pack of 5
Description
- Strep-Tactin™ Sepharose™ columns contains 4% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- Due to the low concentrated agarose, it is particularly suitable for large proteins, especially if the affinity to Strep-Tactin™XT 4Flow™ is too high.
- The wide-meshed polymer allows for the simple attachment of larger proteins to the stationary phase.
- To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
Specifications
| Capacity (Metric) | 0.2 mL |
| For Use With (Application) | Gravity Flow Purification of Strep-Tag Fusion Proteins |
| Type | Gravity Flow Column |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction