Learn More
IBA Lifesciences Strep-Tactin™ Superflow™ High Capacity Column
Gravity flow column to purify Strep-tag™II or Twin-Strep-tag™ fusion proteins
Brand: IBA Lifesciences 2-1209-001
Code : LE
Additional Details : Weight : 0.00100kg
Description
- Strep-Tactin™ Superflow™ high capacity columns contain a 6% agarose coupled with the streptavidin variant Strep-Tactin™ and are applicable for purification of Strep-tag™II or Twin-Strep-tag™ fusion proteins via gravity flow.
- In comparison to Strep-Tactin™ Superflow™, Strep-Tactin™ Superflow™ high capacity has a higher density of immobilized Strep-Tactin™, which leads to a 3- to 5-fold higher yield of purified protein.
- To reduce the waiting period during sample and buffer application, the use of the application aid WET FRED is recommended.
Specifications
1 mL |
For Research Use Only.