missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ C1q Antibody [DyLight 755], Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-11022IR
This item is not returnable.
View return policy
Description
C1q Polyclonal antibody specifically detects C1q in Human samples. It is validated for ELISA,Western Blot
Specifications
| C1q | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Goat | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| DyLight 755 | |
| A chain, complement component 1, q subcomponent, A chain, complement component 1, q subcomponent, alpha polypeptide | |
| Human Complement C1q | |
| 1 mL | |
| Immunology, Innate Immunity | |
| 712 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction