missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Fluorobenzoyl)-propionic acid, 1mg/ml in Methanol, Lipomed

Product Code. 30459852
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30459852

Brand: Lipomed LPMFPA1879FA1LM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

3-(4-Fluorobenzoyl)-propionic acid, 1mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(4-Fluorophenyl)-4-oxobutanoic Acid
CAS 366-77-8
Analyte or Component Names 4-(4-Fluorophenyl)-4-oxobutanoic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H9 F O3
Concentration 1 mg/mL
Synonym Benzenebutanoic acid, 4-fluoro-γ-oxo- (9 CI, ACI), 4-Fluoro-γ-oxobenzenebutanoic acid (ACI), Propionic acid, 3-(p-fluorobenzoyl)- (7 CI, 8 CI), 3-(4-Fluorobenzoyl)propanoic acid, 3-(4-Fluorobenzoyl)propionic acid, 3-(p-Fluorobenzoyl)propionic acid, 4-(4-Fluorophenyl)-4-oxobutanoic acid, 4-(4-Fluorophenyl)-4-oxobutyric acid, 4-Fluorobenzoylpropionic acid, 4-Oxo-4-(p-fluorophenyl)butyric acid, Ba 2833, NSC 408180, β-(4-Fluorobenzoyl)propionic acid, β-(p-Fluorobenzoyl)propionic acid, γ-(4-Fluorophenyl)-γ-oxobutanoic acid
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES OC(=O)CCC(=O)c1ccc(F)cc1
IUPAC Name 4-(4-fluorophenyl)-4-oxobutanoic acid
Solvent or Matrix Methanol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 196.175
Quantity 1 mL
Formula Weight 196.054 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.