missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Voclosporin M1 (IM1-Diol-1) metabolite, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Voclosporin M1 (IM1-Diol-1) metabolite |
| Analyte or Component Names | Voclosporin M1 (IM1-Diol-1) metabolite |
| Shipping Condition | Room Temperature |
| Molecular Formula | C63H113N11O14 |
| InChI Formula | InChI=1S/C63H113N11O14/c1-24-44-59(84)68(17)32-49(77)69(18)45(28-34(2)3)56(81)67-50(38(10)11)62(87)70(19)46(29-35(4)5)55(80)64-41(15)54(79)65-42(16)58(83)71(20)47(30-36(6)7)60(85)72(21)48(31-37(8)9)61(86)73(22)51(39(12)13)63(88)74(23)52(57(82)66-44)53(78)40(14)26-25-27-43(76)33-75/h25,27,34-48,50-53,75-76,78H,24,26,28-33H2,1-23H3,(H,64,80)(H,65,79)(H,66,82)(H,67,81)/b27-25+/t40-,41+,42-,43?,44+,45+,46+,47+,48+,50+,51+,52+,53-/m1/s1 |
| Solution Type | Neat |
| SMILES | C[C@H](C/C=C/C(O)CO)[C@H]([C@]1([H])N(C([C@@H](N(C([C@@H](N(C([C@@H](N(C([C@H](NC([C@@H](NC([C@@H](N(C([C@@H](NC([C@@H](N(C(CN(C([C@@H](NC1=O)CC)=O)C)=O)C)CC(C)C)=O)C(C)C)=O)C)CC(C)C)=O)C)=O)C)=O)C)CC(C)C)=O)C)CC(C)C)=O)C)C(C)C)=O)C)O |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 1248.66 |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction