missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Venlafaxine-d6 N-Oxide, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Venlafaxine-d6 N-Oxide |
| Molecular Formula | C17H21D6NO3 |
| Quantity | 1 mg |
| Synonym | 1-[2-(Dimethyloxidoamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol-d6 |
| InChI Formula | InChI=1S/C17H27NO3/c1-18(2,20)13-16(17(19)11-5-4-6-12-17)14-7-9-15(21-3)10-8-14/h7-10,16,19H,4-6,11-13H2,1-3H3 |
| SMILES | OC1(CC([2H])([2H])C([2H])([2H])C([2H])([2H])C1)C(CN(C)(C)=O)C2=CC=C(OC)C=C2 |
| Molecular Weight (g/mol) | 299.44 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction