missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vaccenic Acid Ethyl-d5 Ester, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Vaccenic Acid Ethyl-d5 Ester |
| Molecular Formula | C20H33D5O2 |
| Quantity | 1 mg |
| Synonym | 11-Octadecenoic Acid Ethyl Ester-d5, (E)-11-Octadecenoic Acid Ethyl Ester-d5, Ethyl Vaccenate-d5 |
| InChI Formula | InChI=1S/C24H35NO4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-24(28)29-25-22(26)20-21-23(25)27/h3-4,6-7,9-10,12-13H,2,5,8,11,14-21H2,1H3/b4-3-,7-6-,10-9-,13-12- |
| SMILES | CC/C=C\C/C=C\C/C=C\C/C=C\CCCCCCC(=O)ON1C(=O)CCC1=O |
| Molecular Weight (g/mol) | 315.55 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction