missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Urolithin A-2,4,7-d3 Major (>85%), TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Urolithin A-2,4,7-d3 |
| Molecular Formula | C13H5D3O4 |
| Quantity | 1 mg |
| Synonym | 3,8-dihydroxy-6H-benzo[c]chromen-6-one-2,4,7-d3 |
| InChI Formula | InChI=1S/C13H8O4/c14-7-1-3-9-10-4-2-8(15)6-12(10)17-13(16)11(9)5-7/h1-6,14-15H/i2D,5D,6D |
| SMILES | O=C1OC2=C([2H])C(O)=C([2H])C=C2C3=C1C([2H])=C(O)C=C3 |
| IUPAC Name | 3,8-dihydroxy-6H-benzo[c]chromen-6-one-2,4,7-d3 |
| Molecular Weight (g/mol) | 231.22 |
| Formula Weight | 231.06 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction