missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Umeclidinium Bromide-d10, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Umeclidinium Bromide-d10 |
| Molecular Formula | C29H24D10BrNO2 |
| Quantity | 1 mg |
| Synonym | 4-(Hydroxydiphenylmethyl)-1-[2-[(phenylmethyl)oxy]ethyl]-1-azabicyclo[2.2.2]octanium bromide-d10, 4-[Hydroxy(diphenyl)methyl]-1-[2-[(phenylmethyl)oxy]ethyl]-1-azoniabicyclo[2.2.2]octane bromide-d10 |
| InChI Formula | InChI=1S/C29H34NO2.BrH/c31-29(26-12-6-2-7-13-26,27-14-8-3-9-15-27)28-16-19-30(20-17-28,21-18-28)22-23-32-24-25-10-4-1-5-11-25;/h1-15,31H,16-24H2;1H/q+1;/p-1/i2D,3D,6D,7D,8D,9D,12D,13D,14D,15D; |
| SMILES | OC(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])(C2=C([2H])C([2H])=C([2H])C([2H])=C2[2H])C34CC[N+](CC4)(CCOCC5=CC=CC=C5)CC3.[Br-] |
| Molecular Weight (g/mol) | 518.55 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction