missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ulipristal-d3, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | Ulipristal-D3 (N-methyl-D3) |
| Molecular Formula | C28 2H3 H32 N O3 |
| Quantity | 1 mg |
| Synonym | (11β)-11-[4-[(Methyl)(methyl-d3)amino]phenyl]-17-hydroxy-19-norpregna-4,9-diene-3,20-dione, 11β-[4-(N-Methyl-N-(methyl-d3)amino)phenyl]-17-hydroxy-19-norpregna-4,9-diene-3,20-dione, Ulipristal-d3, Ulipristal-D3 (N-methyl-D3) |
| InChI Formula | InChI=1S/C28H35NO3/c1-17(30)28(32)14-13-25-23-11-7-19-15-21(31)10-12-22(19)26(23)24(16-27(25,28)2)18-5-8-20(9-6-18)29(3)4/h5-6,8-9,15,23-25,32H,7,10-14,16H2,1-4H3/t23-,24+,25-,27-,28-/m0/s1/i3D3 |
| SMILES | [2H]C([2H])([2H])N(C)c1ccc(cc1)[C@H]2C[C@@]3(C)[C@@H](CC[C@]3(O)C(=O)C)[C@@H]4CCC5=CC(=O)CCC5=C24 |
| IUPAC Name | (8S,11R,13S,14S,17R)-17-acetyl-17-hydroxy-13-methyl-11-[4-[methyl(trideuteriomethyl)amino]phenyl]-1,2,6,7,8,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 436.601 |
| Formula Weight | 436.281 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction