missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Triptonide, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Triptonide |
| CAS | 38647-11-9 |
| Molecular Formula | C20 H22 O6 |
| Quantity | 1 mg |
| Synonym | (-)-Triptonide, NSC 165677, PG 492, Triptonid, 3[3bS-(3bα,4aα,5aR*,6aβ,7aβ,7bα,8aR*,8bβ)]-b,4,4a,7a,7b,8b,9,10-Octahydro-8b-methyl-6a-(1-methylethyl)-trisoxireno[4b,5:6,7:8a,9]phenanthro[1,2-c]furan-1,6(3H,6aH)-dione |
| InChI Formula | InChI=1S/C20H22O6/c1-8(2)18-13(25-18)14-20(26-14)17(3)5-4-9-10(7-23-15(9)21)11(17)6-12-19(20,24-12)16(18)22/h8,11-14H,4-7H2,1-3H3/t11-,12-,13-,14-,17-,18-,19+,20+/m0/s1 |
| SMILES | CC(C)[C@]12O[C@H]1[C@@H]3O[C@@]34[C@@]5(C)CCC6=C(COC6=O)[C@@H]5C[C@@H]7O[C@]47C2=O |
| Molecular Weight (g/mol) | 358.39 |
| Formula Weight | 358.1416 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction