missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trinexapac-d5, TRC
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Chemical Name or Material | Trinexapac-d5 |
| Molecular Formula | C11 D5 H7 O5 |
| Quantity | 1 mg |
| Synonym | 4-(Cyclopropylhydroxymethylene)-3,5-dioxocyclohexanecarboxylic acid-d5, Cimetacarb-d5 |
| InChI Formula | InChI=1S/C11H12O5/c12-7-3-6(11(15)16)4-8(13)9(7)10(14)5-1-2-5/h5-6,14H,1-4H2,(H,15,16)/i1D2,2D2,5D |
| SMILES | [2H]C1([2H])C([2H])([2H])C1([2H])C(=C2C(=O)CC(CC2=O)C(=O)O)O |
| IUPAC Name | 4-[hydroxy-(1,2,2,3,3-pentadeuteriocyclopropyl)methylidene]-3,5-dioxocyclohexane-1-carboxylic acid |
| Molecular Weight (g/mol) | 229.241 |
| Formula Weight | 229.1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction