missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tonalide-d11, TRC
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
Specifications
Specifications
| Chemical Name or Material | Tonalide-d11 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 D11 H15 O |
| Quantity | 1 mg |
| Synonym | 1-(5,6,7,8-Tetrahydro-3,5,5,6,8,8-hexamethyl-2-naphthalenyl)-ethanone-d11, 5',6',7',8'-Tetrahydro-3',5',5',6',8',8'-hexamethyl-2'-acetonaphthone-d11, AHMT (perfume)-d11, Extralide-d11, Fixolide-d11, NSC 19550-d11 |
| InChI Formula | InChI=1S/C18H26O/c1-11-8-16-15(9-14(11)13(3)19)17(4,5)10-12(2)18(16,6)7/h8-9,12H,10H2,1-7H3/i1D3,4D3,5D3,8D,9D |
| SMILES | [2H]c1c(C(=O)C)c(c([2H])c2c1C(CC(C)C2(C)C)(C([2H])([2H])[2H])C([2H])([2H])[2H])C([2H])([2H])[2H] |
| IUPAC Name | 1-[5,8-dideuterio-1,1,2-trimethyl-4,4,7-tris(trideuteriomethyl)tetralin-6-yl]ethanone |
| Molecular Weight (g/mol) | 269.47 |
| Formula Weight | 269.2674 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction