missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tizanidine-d4, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Tizanidine-d4 |
| CAS | 1188331-19-2 |
| Molecular Formula | C9H4D4ClN5S |
| Quantity | 1 mg |
| Synonym | 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl-d4)-2,1,3-benzothiadiazol-4-amine |
| InChI Formula | InChI=1S/C9H8ClN5S/c10-5-1-2-6-8(15-16-14-6)7(5)13-9-11-3-4-12-9/h1-2H,3-4H2,(H2,11,12,13)/i3D2,4D2 |
| SMILES | ClC1=C(NC2=NC([2H])([2H])C([2H])([2H])N2)C3=NSN=C3C=C1 |
| Molecular Weight (g/mol) | 257.74 |
| Formula Weight | 257.044 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction