missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiodiglycolic Acid-d4, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Thiodiglycolic Acid-d4 |
| CAS | 132090-51-8 |
| Molecular Formula | C4 D4 H2 O4 S |
| Quantity | 1 mg |
| Synonym | Acetic-d2 acid','2,2'-thiobis- (9CI), 2,2'-Thiobis[acetic-d2 acid], 2,2'-Thiodiacetic Acid-D4, 2,2'-Thiodiacetic-2,2,2',2'-d4 Acid, Thiodiglycolic Acid-d4 |
| InChI Formula | InChI=1S/C4H6O4S/c5-3(6)1-9-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)/i1D2,2D2 |
| SMILES | [2H]C([2H])(SC([2H])([2H])C(=O)O)C(=O)O |
| IUPAC Name | 2-[carboxy(dideuterio)methyl]sulfanyl-2,2-dideuterioacetic acid |
| Molecular Weight (g/mol) | 154.178 |
| Formula Weight | 154.024 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction