missing translation for 'onlineSavingsMsg'
Learn More

Sofosbuvir-d6, MedChemExpress

Product Code. 16451469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16451469 1 mg 1mg
16458877 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16451469 Supplier Medchem Express Supplier No. HY15005S1
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Sofosbuvir D6 (PSI-7977 D6) is the deuterium labeled Sofosbuvir. Sofosbuvir (PSI-7977) is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.

Sofosbuvir D6 (PSI-7977 D6) is the deuterium labeled Sofosbuvir. Sofosbuvir (PSI-7977) is an active inhibitor of HCV RNA replication in the HCV replicon assay, demonstrates potent anti-hepatitis C virus (HCV) activity.

Specifications

Chemical Name or Material Sofosbuvir-d6
CAS 1868135-06-1
Purity Grade Notes Research
Physical Form Solid
Molecular Formula C22H23D6FN3O9P
Color White
Synonym PSI-7977-d6 GS-7977-d6
SMILES O=C1N([C@H]2[C@]([C@H](O)[C@@H](CO[P@](OC3=CC=CC=C3)(N[C@@H](C)C(OC(C([2H])([2H])[2H])C([2H])([2H])[2H])=O)=O)O2)(C)F)C=CC(N1)=O
Molecular Weight (g/mol) 535.49
Quantity 1 mg
Formula Weight 535.49
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.