missing translation for 'onlineSavingsMsg'
Learn More
Learn More
rac-Vigabatrin-13C,d2 (Major), TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | rac-Vigabatrin-13C,d2 |
| CAS | 1330171-61-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C513CH9D2NO2 |
| Quantity | 1 mg |
| Synonym | (+/-)-4-Amino-5-hexenoic Acid-13C,d2, (+/-)-γ-Vinyl GABA-13C,d2, GVG-13C,d2, MDL 71754-13C,d2, RMI 71754-13C,d2, Sabril-13C,d2, Vigabatrin-13C,d2, γ-Vinyl-GABA-13C,d2, γ-Vinyl-γ-aminobutyric Acid-13C,d2 |
| InChI Formula | InChI=1S/C6H11NO2/c1-2-5(7)3-4-6(8)9/h2,5H,1,3-4,7H2,(H,8,9)/i1+1D2 |
| SMILES | NC(/C=[13C]([2H])/[2H])CCC(O)=O |
| Molecular Weight (g/mol) | 132.16 |
| Percent Purity | >95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction