Learn More
Abcam 1MG Potent, selective, ATP competitive JNK inhibitor. Cell-permeable.
Brand:  Abcam ab145194-1mg
Encompass
1MG Potent, selective, ATP competitive JNK inhibitor. Cell-permeable.
Product Code. 16257194
  • £120.00 / 1mg