missing translation for 'onlineSavingsMsg'
Learn More

Oligomycin A, MedChemExpress

Product Code. 16401629
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16401629

Brand: Medchem Express HY16589

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Oligomycin A (MCH 32), created by Streptomyces, acts as a mitochondrial F0F1-ATPase inhibitor, with a Ki of 1 μM; Oligomycin A shows anti-fungal activity.

Oligomycin A (MCH 32), created by Streptomyces, acts as a mitochondrial F0F1-ATPase inhibitor, with a Ki of 1 μM

Specifications

Chemical Name or Material Oligomycin A
Physical Form Powder
CAS 579-13-5
Color White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C45H74O11
For Use With (Application) Cancer-programmed cell death
Synonym MCH 32
Solubility Information DMSO : 100 mg/mL (126.41 mM; Need ultrasonic)
SMILES O=C(/C=C/[C@H](C)[C@@H](O)[C@H](C)C([C@H](C)[C@@H](O)[C@@H]1C)=O)O[C@@]([C@@H]2C)([H])[C@H](C)[C@@]3(CC[C@H](C)[C@H](C[C@H](O)C)O3)O[C@@]2([H])CC[C@@H](CC)/C=C/C=C/C[C@@H](C)[C@H](O)[C@@](C)(O)C1=O
Molecular Weight (g/mol) 791.06
Formula Weight 791.06
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.