missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Vanillylnonanamide-d3, TRC
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
Specifications
Specifications
| Chemical Name or Material | N-Vanillylnonanamide-d3 |
| Molecular Formula | C17H24D3NO3 |
| Quantity | 1 mg |
| Synonym | N-[(4-Hydroxy-3-methoxyphenyl)methyl]nonanamide-d3, 8-Nordihydrocapsaicin-d3, AH 23491X-d3, N-(4-Hydroxy-3-methoxybenzyl)nonamide-d3, N-Vanillylnonamide-d3, N-Vanillylpelargonamide-d3, NSC 172795-d3, Nonanoic Acid Vanillylamide-d3, Nonanoylvanillyl Amide-d3, Nonivamide-d3, Nonylic Acid Vanillylamide-d3, Nonylic Vanillylamide-d3, Pelargonic Acid Vanillylamide-d3, Pseudocapsaicin-d3, Vanillyl n-Nonylamide-d3, Vanillyl Pelargonic Amide-d3 |
| InChI Formula | InChI=1S/C18H29NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h10-12,14,20H,4-9,13H2,1-3H3,(H,19,21)/i3D3 |
| SMILES | OC1=CC=C(CNC(CCCCCCCC)=O)C=C1OC([2H])([2H])[2H] |
| Molecular Weight (g/mol) | 296.42 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction