missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MuSK Antibody (Argenx patent anti-MuSK) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28736-1mg
This item is not returnable.
View return policy
Description
MuSK Monoclonal antibody specifically detects MuSK in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| MuSK | |
| Monoclonal | |
| Unconjugated | |
| O15146 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Argenx patent anti-MuSK | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| EC 2.7.10, EC 2.7.10.1, MGC126323, MGC126324, muscle, skeletal, receptor tyrosine kinase, MuSK, skeletal receptor tyrosine-protein kinase | |
| MUSK | |
| 1 mg | |
| Protein Kinase, Signal Transduction | |
| 4593 | |
| Human | |
| Purified |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido