Learn More
DiscoverX Calcitonin, human
Product Code. 17372389
Encompass
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. Quantity unitSize
17372389 1 mg 1mg
1 options
Product Code. 17372389 Supplier DiscoverX Supplier No. 921054
This item requires a price update prior to being ordered.
Request a Price for this item here.
Calcitonin, human is an agonist for CALCR. Provided as 1mg of dry powder alongside reconstitution buffer.
Calcitonin is a polypeptide hormone produced in the thyroid by parafollicular cells (C-Cells). It reduces blood calcium levels in opposition to the effects of parathyroid hormone. It participates in both calcium and phosphorus metabolism, stimulates osteoblast activity, and inhibits intestinal calcium absorption, osteoclast activity, and renal tubular cell calcium absorption. Calcitonin levels in the blood are dictated by blood calcium levels. Acts as an agonist for CALCR. Provided as 1mg of dry powder alongside reconstitution buffer.
Specifications
Product Type Control Ligands
Quantity 1 mg