missing translation for 'onlineSavingsMsg'
Learn More

Beta-Pseudouridine-13C, 15N2, TRC

Product Code. 30005216
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005216 1 mg 1mg
30007754 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30005216 Supplier Toronto Research Chemicals Supplier No. TRCP8396071MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C813CH1215N2O6
Molecular Weight (g/mol) 247.18
Synonym 5-β-D-Ribofuranosyl-2,4(1H,3H)-Pyrimidinedione-13C, 15N2, 5-(β-D-Ribofuranosyl)uracil-13C, 15N2, 5-Ribosyluracil-13C, 15N2, NSC 162405-13C, 15N2, Pseudouridine C, β-D-Pseudouridine-13C, 15N2, Pseudouridine-13C, 15N2, ψ-Uridine-13C, 15N2;
IUPAC Name 5-((2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione-2-13C-1,3-15N2
SMILES O[C@@H]1[C@H](O)[C@@H](CO)O[C@H]1C(C([15NH]2)=O)=C[15NH][13C]2=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C9H12N2O6/c12-2-4-5(13)6(14)7(17-4)3-1-10-9(16)11-8(3)15/h1,4-7,12-14H,2H2,(H2,10,11,15,16)/t4-,5-,6-,7+/m1/s1/i9+1,10+1,11+1
Formula Weight 247.07
Percent Purity >95%
Chemical Name or Material b-Pseudouridine-13C, 15N2
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.