missing translation for 'onlineSavingsMsg'
Learn More

25-Deacetyl-21-acetyl Rifampicin, TRC

Product Code. 30010806
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010806 1 mg 1mg
30007854 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010806 Supplier Toronto Research Chemicals Supplier No. TRCD1982051MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1416773-22-2
Molecular Formula C43H58N4O12
Molecular Weight (g/mol) 822.94
Synonym 25-O-Deacetyl-3-[[(4-methyl-1-piperazinyl)imino]methyl]rifamycin 21-acetate, 25-O-Deacetylrifampicin 21-acetate, 25-Deacetyl-21-acetyl rifampicin, 25-Desacetyl-21-acetylrifampicin, Rifampicin impurity 2
IUPAC Name [(7R,9E,11R,12S,13R,14R,15S,16R,17S,18R,19E,21E)-2,13,15,27,29-pentahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-26-[(E)-(4-methylpiperazin-1-yl)iminomethyl]-6,23-dioxo-8,30-dioxa-24-azatetracyclo[23.3.1.14,7.05,28]triaconta-1(29),2,4,9,19,21,25,27-octaen-17-yl] acetate
SMILES CO[C@H]1\C=C\O[C@@]2(C)Oc3c(C)c(O)c4c(O)c(NC(=O)\C(=C/C=C/[C@H](C)[C@H](OC(=O)C)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](O)[C@@H]1C)\C)c(\C=N\N5CCN(C)CC5)c(O)c4c3C2=O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C43H58N4O12/c1-21-12-11-13-22(2)42(55)45-33-28(20-44-47-17-15-46(9)16-18-47)37(52)30-31(38(33)53)36(51)26(6)40-32(30)41(54)43(8,59-40)57-19-14-29(56-10)23(3)34(49)24(4)35(50)25(5)39(21)58-27(7)48/h11-14,19-21,23-25,29,34-35,39,49-53H,15-18H2,1-10H3,(H,45,55)/b12-11+,19-14+,22-13+,44-20+/t21-,23-,24-,25-,29-,34+,35+,39+,43-/m1/s1
Formula Weight 822.4051
Chemical Name or Material 25-Deacetyl-21-acetyl Rifampicin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.