missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reagecon 1LT Buffer Standard pH 10.00 20oC ISO 17034 CRM Certified Reference Material
Brand: Reagecon CRM1100
1LT Buffer Standard pH 10.00 20oC ISO 17034 CRM Certified Reference Material
This item is not returnable.
View return policy
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- BUFFER STANDARD pH 10.00 at 20°C ISO 17034 CERTIFIED REFERENCE MATERIAL
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : boric acid, disodiumsalt
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction