missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-3-boronic acid pinacol ester, 95%

Product Code. 11446560
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11446560 5 g 5g
11436560 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446560 Supplier Thermo Scientific Alfa Aesar Supplier No. H27619.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 844501-71-9 | C9H15BN2O2 | 194.041 g/mol

1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1H-Pyrazole-3-boronic acid pinacol ester is used as an intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions. Keep container tightly closed. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 844501-71-9
Molecular Formula C9H15BN2O2
Molecular Weight (g/mol) 194.041
MDL Number MFCD07368044
InChI Key KWLOIDOKWUESNM-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-3-boronic acid pinacol ester, pyrazole-3-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-pyrazole, 3-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1h-pyrazole-5-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxabolone-pyrrazole, 1h-pyrazol-3-yl-boronic acid pinacol ester, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolone-pyrrazole
PubChem CID 21931539
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC=NN2

Specifications

Melting Point 105°C to 108°C
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 194.04
Percent Purity 95%
Chemical Name or Material 1H-Pyrazole-3-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.