missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-3-boronic acid hydrate, 95%

Product Code. 11372339
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11372339 5 g 5g
11362339 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372339 Supplier Thermo Scientific Alfa Aesar Supplier No. H27286.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 376584-63-3 | C3H5BN2O2 | 111.90 g/mol

Significantly used as synthetic intermediates. They are widely used in the production of optics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Significantly used as synthetic intermediates. They are widely used in the production of optics.

Solubility
Soluble in water.

Notes
Refrigerated (2-8°C). Moisture sensitive. Store under inert gas. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 376584-63-3
Molecular Formula C3H5BN2O2
Molecular Weight (g/mol) 111.90
MDL Number MFCD07368247 MFCD02020768
InChI Key NEUWPDLMDVINSN-UHFFFAOYSA-N
Synonym 1h-pyrazol-3-yl boronic acid, pyrazole-3-boronic acid, 1h-pyrazol-3-ylboronic acid, 1h-pyrazole-3-boronic acid, 1h-pyrazole-5-boronic acid, 1h-pyrazol-5-yl boronic acid, 1h-pyrazol-5-yl-5-boronic acid, pyrazol-3-ylboronic acid, 2h-pyrazol-3-ylboronic acid
PubChem CID 11251979
IUPAC Name 1H-pyrazol-5-ylboronic acid
SMILES OB(O)C1=NNC=C1

Specifications

Melting Point 85°C to 88°C
Quantity 5 g
Formula Weight 111.9
Percent Purity 95%
Chemical Name or Material 1H-Pyrazole-3-boronic acid hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.