missing translation for 'onlineSavingsMsg'
Learn More

1H-Pyrazole-3,5-dicarboxylic acid monohydrate, 98%

Product Code. 11326976
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11326976 5 g 5g
11336976 25 g 25g
11346976 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11326976 Supplier Thermo Scientific Alfa Aesar Supplier No. L06731.06
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 303180-11-2 | C5H6N2O5 | 174.112 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 303180-11-2
Molecular Formula C5H6N2O5
Molecular Weight (g/mol) 174.112
MDL Number MFCD00149323
InChI Key GLINCONFUZIMCN-UHFFFAOYSA-N
Synonym 1h-pyrazole-3,5-dicarboxylic acid hydrate, 3,5-pyrazoledicarboxylic acid monohydrate, 1h-pyrazole-3,5-dicarboxylic acid monohydrate, pyrazole-3,5-dicarboxylic acid, hydrate, acmc-1aezk, 3,5-pyrazoledicarboxylic acid hydrate, 1h-pyrazole-3,5-dicarboxylicacidmonohydrate, 1h-pyrazole-3,5-dicarboxylic acid; hydrate, 1h-pyrazole-3,5-dicarboxylic acid hydrate 1:1
PubChem CID 2723723
IUPAC Name 1H-pyrazole-3,5-dicarboxylic acid;hydrate
SMILES C1=C(NN=C1C(=O)O)C(=O)O.O

Specifications

Melting Point ∽294°C (decomposition)
Quantity 5 g
Beilstein 147821
Formula Weight 174.11 (156.09 Anhydrous)
Percent Purity 98%
Chemical Name or Material 1H-Pyrazole-3,5-dicarboxylic acid monohydrate
compliance-icons
Product Identifier
  • 1H-Pyrazole-3,5-dicarboxylic acid monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 1H-Pyrazole-3,5-dicarboxylic acid monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.