missing translation for 'onlineSavingsMsg'
Learn More

1H-Perfluorohexane, 97%, Thermo Scientific Chemicals

Product Code. 11479686
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479686 5 g 5g
11489686 25 g 25g
11499686 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11479686 Supplier Thermo Scientific Alfa Aesar Supplier No. B20226.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 355-37-3
Molecular Formula C6HF13
Molecular Weight (g/mol) 320.05
MDL Number MFCD00142632
InChI Key XJSRKJAHJGCPGC-UHFFFAOYSA-N
Synonym 1h-perfluorohexane, 1h-tridecafluorohexane, hexane, tridecafluoro, trideca-1,1,1,2,2,3,3,4,4,5,5,6,6-fluorohexane, hexane, 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluoro, tridecafluorohexane, acmc-209tqg, #
PubChem CID 67730
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6-tridecafluorohexane
SMILES FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Density 1.6843
Boiling Point 70°C to 71°C
Flash Point None
Linear Formula CF3(CF2)5H
Refractive Index <1.300
Quantity 5 g
Beilstein 1799000
Formula Weight 320.05
Percent Purity 97%
Chemical Name or Material 1H-Perfluorohexane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.