missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-5-carboxamide, TRC

Product Code. 30401075
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401075 10 mg 10mg
30390507 5 mg -
30389917 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401075 Supplier Toronto Research Chemicals Supplier No. TRCI61550010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1670-87-7
Molecular Formula C9H8N2O
Molecular Weight (g/mol) 160.17
Synonym Indole-5-carboxamide, 5-(Aminocarbonyl)-1H-indole, 5-Carbamoylindole, SD 169
SMILES NC(=O)c1ccc2[nH]ccc2c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H8N2O/c10-9(12)7-1-2-8-6(5-7)3-4-11-8/h1-5,11H,(H2,10,12)
Formula Weight 160.0637
Chemical Name or Material 1H-Indole-5-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.