missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-3-carbonitrile, 97%, Maybridge™

Product Code. 10041724
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041724 1 g Amber Glass Bottle 1g
10051004 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041724 Supplier Thermo Scientific Maybridge Supplier No. RF01264DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5457-28-3
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.16
MDL Number MFCD00022717
InChI Key CHIFTAQVXHNVRW-UHFFFAOYSA-N
Synonym 3-cyanoindole, indole-3-carbonitrile, 3-indolecarbonitrile, chembl46724, zlchem 360, 3-cyano-1h-indole, pubchem7325, acmc-209lhi, 1h-indol-3-yl cyanide
PubChem CID 230282
IUPAC Name 1H-indole-3-carbonitrile
SMILES N#CC1=CNC2=CC=CC=C12

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 142.16
Chemical Name or Material 1H-Indole-3-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.