missing translation for 'onlineSavingsMsg'
Learn More

(1H-Indol-3-yl)methanamine Hydrochloride, TRC

Product Code. 30393892
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393892 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393892 Supplier Toronto Research Chemicals Supplier No. TRCI628200250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1266692-14-1
Molecular Formula C9H10N2 . HCl
Quantity 250 mg
Synonym 1H-Indol-3-ylmethanamine hydrochloride
InChI Formula InChI=1S/C9H10N2.ClH/c10-5-7-6-11-9-4-2-1-3-8(7)9;/h1-4,6,11H,5,10H2;1H
SMILES C1=CC=C2C(=C1)C(=CN2)CN.Cl
IUPAC Name (1H-indol-3-yl)methanamine hydrochloride
Molecular Weight (g/mol) 182.65
Formula Weight 182.54
Chemical Name or Material (1H-Indol-3-yl)methanamine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.