missing translation for 'onlineSavingsMsg'
Learn More

1H-Indol-3-amine Hydrochloride, TRC

Product Code. 30408029
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408029 50 mg -
30396766 250 mg 250mg
30404740 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30408029 Supplier Toronto Research Chemicals Supplier No. TRCI62822350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57778-93-5
Molecular Formula C8H8N2 . HCl
Molecular Weight (g/mol) 168.62
Synonym 3-Aminoindole HCl
IUPAC Name 1H-indol-3-amine;hydrochloride
SMILES Cl.Nc1c[nH]c2ccccc12

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H8N2.ClH/c9-7-5-10-8-4-2-1-3-6(7)8;/h1-5,10H,9H2;1H
Formula Weight 168.0454
Chemical Name or Material 1H-Indol-3-amine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.