missing translation for 'onlineSavingsMsg'
Learn More

1H-Imidazole-5-acetic Acid Hydrochloride, TRC

Product Code. 30009737
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
10g
2.5g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009737 2.5 g 2.5g
30008786 10 g 10g
30006472 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009737 Supplier Toronto Research Chemicals Supplier No. TRCI3856052.5G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3251-69-2
Molecular Formula C5 H6 N2 O2 . Cl H
Molecular Weight (g/mol) 162.57
Synonym Imidazoleacetic Acid Hydrochloride, (1H-Imidazol-4-yl)acetic Acid Hydrochloride, (1H-Imidazol-5-yl)acetic Acid Hydrochloride, 1H-Imidazole-4-acetic Acid Hydrochloride, 1H-Imidazole-5-acetic Acid Hydrochloride, Imidazol-4-acetic Acid Hydrochloride, Imidazole-4-acetic Acid Hydrochloride
IUPAC Name 2-(1H-imidazol-4-yl)acetic acid;hydrochloride
SMILES Cl.OC(=O)Cc1c[nH]cn1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C5H6N2O2.ClH/c8-5(9)1-4-2-6-3-7-4;/h2-3H,1H2,(H,6,7)(H,8,9);1H
Formula Weight 162.0196
Chemical Name or Material 1H-Imidazole-5-acetic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.