missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-5-carboxylic Acid, TRC

Product Code. 30345174
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30345174 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30345174

Brand: Toronto Research Chemicals TRCB19702010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1H-Benzimidazole-5-carboxylic Acid
CAS 15788-16-6
Molecular Formula C8H6N2O2
Quantity 10 g
Synonym 1H-Benzimidazole-6-carboxylic Acid, 5-Benzimidazolecarboxylic Acid, 3H-Benzimidazole-5-carboxylic Acid, 3H-Benzo[d]imidazole-5-carboxylic Acid, 5-Carboxybenzimidazole, Benzimidazole-6-carboxylic Acid
InChI Formula InChI=1S/C8H6N2O2/c11-8(12)5-1-2-6-7(3-5)10-4-9-6/h1-4H,(H,9,10)(H,11,12)
SMILES OC(=O)c1ccc2[nH]cnc2c1
IUPAC Name 1H-benzimidazole-5-carboxylic acid
Molecular Weight (g/mol) 162.15
Formula Weight 162.0429
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.