missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-5-carboxylic acid, 98%

Product Code. 10422831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10422831 25 g 25g
10427952 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10422831 Supplier Thermo Scientific Acros Supplier No. 290870250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15788-16-6 | C8H6N2O2 | 162.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15788-16-6
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00011555
InChI Key COYPLDIXZODDDL-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carboxylic acid, 5-benzimidazolecarboxylic acid, benzimidazole-5-carboxylic acid, 1h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-6-carboxylic acid, 5-carboxybenzimidazole, 1h-1,3-benzodiazole-5-carboxylic acid, 3h-benzo d imidazole-5-carboxylic acid, 1h-benzimidazole-5-carboxylicacid, 1h-1,3-benzodiazole-6-carboxylic acid
PubChem CID 459456
IUPAC Name 3H-benzimidazole-5-carboxylic acid
SMILES OC(=O)C1=CC=C2N=CNC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point >300°C
Color Brown-Gray to Gray-Green
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 25 g
Formula Weight 162.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1H-Benzimidazole-5-carboxylic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1H-Benzimidazole-5-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.