missing translation for 'onlineSavingsMsg'
Learn More

1H-Benzimidazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10055483
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10055483 5 g Amber Glass Bottle 5g
10794255 250 mg Amber Glass Bottle 250mg
10092314 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10055483 Supplier Thermo Scientific Maybridge Supplier No. CC45904DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kemiske identifikatorer

CAS 58442-17-4
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.149
InChI Key ALCHVVTYAHQOFY-UHFFFAOYSA-N
Synonym 1h-benzimidazole-5-carbaldehyde, 1h-benzimidazole-5-carboxaldehyde, 1h-benzo d imidazole-5-carbaldehyde, benzimidazole-5-aldehyde, 1h-1,3-benzodiazole-5-carbaldehyde, 1h-benzimidazole-6-carboxaldehyde, benzimidazole-5-carboxaldehyde, 1h-benzoimidazole-5-carbaldehyde, 3h-benzoimidazole-5-carbaldehyde, pubchem9866
PubChem CID 10012038
IUPAC Name 3H-benzimidazole-5-carbaldehyde
SMILES C1=CC2=C(C=C1C=O)NC=N2

Tekniske data

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 146.15
Percent Purity 97%
Chemical Name or Material 1H-Benzimidazole-5-carbaldehyde
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.