Learn More
Thermo Scientific Maybridge 1H-Benzimidazol-5-ylmethanol, 1g, 95%
Brand: Thermo Scientific Maybridge CC45909DA
1H-Benzimidazol-5-ylmethanol, 1g, 95%
Code : 31
Additional Details : CAS Number : 106429-29-2 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III