missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluorooctyl acrylate, 97%, stab. with 50ppm 4-methoxyphenol, Thermo Scientific™

Product Code. 11359275
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11359275

Brand: Thermo Scientific Alfa Aesar L16603.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-98-2
Molecular Formula C11H5F15O2
Molecular Weight (g/mol) 454.135
MDL Number MFCD00039248
InChI Key YSQGYEYXKXGAQA-UHFFFAOYSA-N
Synonym 1h,1h-pentadecafluorooctyl acrylate, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl acrylate, 1h,1h-perfluorooctyl acrylate, acrylic acid 1h,1h-pentadecafluoro-n-octyl ester, 2-propenoic acid, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl ester, 1h,1h-perfluoro-n-octyl acrylate, 1h,1h-pentadecafluoro-n-octyl acrylate, 1,1-dihydroperfluorooctyl acrylate, 1,1-dyhydroperfluorooctyl acrylate, acrylic acid 1h,1h-perfluorooctyl ester
PubChem CID 67551
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl prop-2-enoate
SMILES C=CC(=O)OCC(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.403
Boiling Point 112°C to 115°C (60 mmHg)
Flash Point 91°C (195°F)
Linear Formula CF3(CF2)6CH2O2CCH=CH2
Refractive Index 1.3275
Quantity 1 g
Beilstein 1809801
Formula Weight 454.14
Percent Purity 97%
Chemical Name or Material 1H,1H-Perfluorooctyl acrylate, stabilized with 50ppm 4-methoxyphenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.