missing translation for 'onlineSavingsMsg'
Learn More

1H,1H-Perfluoro-1-octanol, 98%

Product Code. 10116923 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10116923 25 g Glass bottle 25g
10217650 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 10116923 Supplier Thermo Scientific Acros Supplier No. 277260250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

CAS: 307-30-2 | C8H3F15O | 400.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 307-30-2
Molecular Formula C8H3F15O
Molecular Weight (g/mol) 400.08
MDL Number MFCD00004675
InChI Key PJDOLCGOTSNFJM-UHFFFAOYSA-N
Synonym 1h,1h-pentadecafluoro-1-octanol, 1h,1h-perfluoro-1-octanol, 1h,1h-perfluorooctanol, pentadecafluorooctyl alcohol, unii-9x5w7fq62r, 1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro, pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluoro-1-octanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctyl alcohol, perfluorooctanol
PubChem CID 67543
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-pentadecafluorooctan-1-ol
SMILES C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 43.0°C to 47.0°C
Color White
Boiling Point 100°C to 102°C (100.0 hPa)
Flash Point >150°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3(CF2)6CH2OH
Quantity 25 g
Beilstein 01, IV, 1766
Solubility Information Solubility in water: insoluble
Formula Weight 400.08
Percent Purity 98%
Physical Form Crystalline Mass
Chemical Name or Material 1H, 1H-Perfluoro-1-octanol
Show More Show Less
compliance-icons
Product Identifier
  • 1H,1H-Perfluoro-1-octanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.