missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,9H-Perfluoro-1-nonanol, 97%, Thermo Scientific Chemicals

Product Code. p-7025748
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11449367

Brand: Thermo Scientific Alfa Aesar B21407.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 376-18-1
Molecular Formula C9H4F16O
Molecular Weight (g/mol) 432.104
MDL Number MFCD00039629
InChI Key MSXVQELLSMPBFD-UHFFFAOYSA-N
Synonym 1h,1h,9h-hexadecafluoro-1-nonanol, hexadecafluoro-1-nonanol, 1-nonanol, hexadecafluoro, 1-nonanol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluoro, 1-nonanol, 1h,1h,9h-hexadecafluoro, 1h,1h,9h-hexadecafluoro-1-hydroxynonane, 1h,1h,9h-perfluoro-1-nonanol, 1h,1h,9h-hexadecafluorononan-1-ol, omega-h-hexadekafluornonanol-1 german, 1h,1h,9h-hexadecafluorononanol
PubChem CID 9779
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-hexadecafluorononan-1-ol
SMILES C(C(C(C(C(C(C(C(C(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)O

Especificaciones

Melting Point 55°C to 60°C
Boiling Point 157°C to 158°C (200 mmHg)
Quantity 5 g
Beilstein 1808149
Formula Weight 432.11
Percent Purity 97%
Chemical Name or Material 1H,1H,9H-Perfluoro-1-nonanol

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado