missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-octene, TRC

Product Code. 30136908
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136908 500 mg 500mg
30040385 100 mg 100mg
30040516 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136908 Supplier Toronto Research Chemicals Supplier No. TRCP999228500MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25291-17-2
Molecular Formula C8H3F13
Molecular Weight (g/mol) 346.09
Synonym 1-Octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro- (8CI, 9CI, ACI), 3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octene (ACI), (Perfluorohexyl)ethene, (Perfluorohexyl)ethylene, 1,1,2-Trihydroperfluoro-1-octene, 1H,1H,2H,2H-Perfluorooct-1-ene, 1H,1H,2H-Perfluoro-1-octene, 1H,1H,2H-Tridecafluoro-1-octene, C 60LF, Cheminox PFHE, ET 600, ET 600 (fluorocompound), F 1620, TE 6
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C8H3F13/c1-2-3(9,10)4(11,12)5(13,14)6(15,16)7(17,18)8(19,20)21/h2H,1H2
Formula Weight 346.0027
Chemical Name or Material 1H,1H,2H-Perfluoro-1-octene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.