missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-octene, 99%

Product Code. 11499956
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499956 25 g 25g
11489956 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499956 Supplier Thermo Scientific Alfa Aesar Supplier No. B20920.14
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 25291-17-2 | C8H3F13 | 346.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25291-17-2
Molecular Formula C8H3F13
Molecular Weight (g/mol) 346.091
MDL Number MFCD00039249
InChI Key FYQFWFHDPNXORA-UHFFFAOYSA-N
Synonym perfluorohexyl ethylene, 1h,1h,2h-perfluoro-1-octene, tridecafluorohexyl ethylene, 1-octene, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro, unii-j4kbi9s48d, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-1-octene, j4kbi9s48d, perfluorohexylethylene, acmc-209gix, c6f13ch=ch2
PubChem CID 91384
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooct-1-ene
SMILES C=CC(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.58
Boiling Point 106°C to 107°C
Flash Point 20°C (202°F)
Refractive Index 1.296
Quantity 25 g
Beilstein 2060994
Formula Weight 346.09
Percent Purity 99%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-octene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.