missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-hexene, 97%

Product Code. 11393776
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11393776 25 g 25g
11383776 5 g 5g
11303786 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11393776 Supplier Thermo Scientific Alfa Aesar Supplier No. L16834.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 19430-93-4 | C6H3F9 | 246.076 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19430-93-4
Molecular Formula C6H3F9
Molecular Weight (g/mol) 246.076
MDL Number MFCD00042338
InChI Key GVEUEBXMTMZVSD-UHFFFAOYSA-N
Synonym perfluorobutyl ethylene, 3,3,4,4,5,5,6,6,6-nonafluoro-1-hexene, perfluorobutylethylene, 1h,1h,2h-perfluoro-1-hexene, 1h,1h,2h-perfluorohexene, perfluorobutyl ethene, 1-hexene, 3,3,4,4,5,5,6,6,6-nonafluoro, 3,3,4,4,5,5,6,6,6-nonafluorohexene, perfluoro-n-butyl ethylene, 1h,1h,2h-perfluorohex-1-ene
PubChem CID 88054
IUPAC Name 3,3,4,4,5,5,6,6,6-nonafluorohex-1-ene
SMILES C=CC(C(C(C(F)(F)F)(F)F)(F)F)(F)F

Specifications

Density 1.457
Boiling Point 59°C to 60°C
Flash Point None
Linear Formula CF3(CF2)3CH=CH2
Refractive Index 1.289
Quantity 25 g
Beilstein 1819716
Formula Weight 246.08
Percent Purity 97%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-hexene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.