missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H,1H,2H-Perfluoro-1-dodecene, TRC
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
Specifications
Specifications
| Chemical Name or Material | 1H,1H,2H-Perfluoro-1-dodecene |
| CAS | 30389-25-4 |
| Purity Grade Notes | GC |
| Molecular Formula | C12 H3 F21 |
| Quantity | 500 mg |
| Synonym | 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluoro-1-dodecene, (Perfluorodecyl)ethylene, 1,1,2-Trihydroperfluoro-1-dodecene, 1,1,2-Trihydroperfluorododecene, 1H,1H,2H-Perfluoro-1-dodecene, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluorododecene, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Henicosafluorododec-1-ene |
| InChI Formula | InChI=1S/C12H3F21/c1-2-3(13,14)4(15,16)5(17,18)6(19,20)7(21,22)8(23,24)9(25,26)10(27,28)11(29,30)12(31,32)33/h2H,1H2 |
| SMILES | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C |
| IUPAC Name | 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-henicosafluorododec-1-ene |
| Molecular Weight (g/mol) | 546.119 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction