missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H-Perfluoro-1-decene, 99%, Thermo Scientific™

Product Code. 11355299
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355299 25 g 25g
11345299 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355299 Supplier Thermo Scientific Alfa Aesar Supplier No. B20231.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1H,1H,2H-Perfluoro-1-decene can be used to prepare perfluorononan-1-oic acid. Appliance Paint, Boat Paint, Buidling. It con be used as monomer and key intermediate of fluoro silicon surfactant.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21652-58-4
Molecular Formula C10H3F17
Molecular Weight (g/mol) 446.11
MDL Number MFCD00039246
InChI Key NKAMGQZDVMQEJL-UHFFFAOYSA-N
Synonym 1h,1h,2h-perfluoro-1-decene, perfluoro-n-octyl ethylene, perfluorooctyl ethylene, tridecafluoro 1-decene, 1h,1h,2h-perfluorodec-1-ene, 1h,1h,2h-heptadecafluoro-1-decene, perfluoroctyl ethylene, cf3 cf2 7ch=ch2, 1-heptadecafluorooctyl ethene
PubChem CID 88990
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene
SMILES FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C=C

Specifications

Density 1.68
Boiling Point 146°C to 147°C
Flash Point 125°C (257°F)
Linear Formula CF3(CF2)7CH=CH2
Refractive Index 1.3033
Quantity 25 g
Beilstein 2066558
Solubility Information Insoluble in water.
Formula Weight 446.11
Percent Purity 99%
Chemical Name or Material 1H,1H,2H-Perfluoro-1-decene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.