missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane, 97%

Product Code. 11319285
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11319285 25 g 25g
11309285 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11319285 Supplier Thermo Scientific Alfa Aesar Supplier No. L16605.14
 Restricted Item
Request Bulk or Custom Formats

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 73609-36-6 | C9H7Cl2F13Si | 461.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73609-36-6
Molecular Formula C9H7Cl2F13Si
Molecular Weight (g/mol) 461.12
MDL Number MFCD00042360
InChI Key VBDMVWQNRXVEGC-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyl methyldichlorosilane, 1h,1h,2h,2h-perfluorooctylmethyldichlorosilane, dichloro methyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl methyldichlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-methyldichlorosilane, dichloro-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, bis chloranyl-methyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecakis fluoranyl octyl silane, silane, dichloromethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl, 2-tridecafluorohexyl ethyldichloro methyl silane
PubChem CID 123573
IUPAC Name dichloro-methyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C[Si](Cl)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Boiling Point 189°C to 190°C
Quantity 25 g
Flash Point 51°C (123°F)
Refractive Index 1.35
Sensitivity Moisture sensitive
Density 1.55
UN Number UN2920
Beilstein 6153667
Formula Weight 461.13
Percent Purity 97%
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.