missing translation for 'onlineSavingsMsg'
Learn More

1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane, 97%

Product Code. 11435300
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435300 2 g 2g
11445300 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11435300 Supplier Thermo Scientific Alfa Aesar Supplier No. 044543.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 102488-47-1 | C10H10ClF13Si | 440.70 g/mol

1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane, can be used to produce dimethyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-octyl)-silanol. This reaction will need reagent H2O, (NH4)2CO3, NaCl and solvent diethyl ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102488-47-1
Molecular Formula C10H10ClF13Si
Molecular Weight (g/mol) 440.70
MDL Number MFCD00042359
InChI Key AKYGPHVLITVSJE-UHFFFAOYSA-N
Synonym 1h,1h,2h,2h-perfluorooctyldimethylchlorosilane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, tridecafluoro-1,1,2,2-tetrahydrooctyl dimethylchlorosilane, 2-tridecafluorohexyl ethyldimethylchlorosilane, tridecafluoro-1,1,2,2-tetrahydrooctyl-1-dimethylchlorosilane, 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyldimethylchlorosilane, chloro-dimethyl-3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl silane, silane, chlorodimethyl 3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl
PubChem CID 145382
IUPAC Name chloro-dimethyl-(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluorooctyl)silane
SMILES C[Si](C)(Cl)CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Specifications

Boiling Point 189°C to 191°C
Color Colorless
Physical Form Liquid
Quantity 2 g
UN Number UN2986
Beilstein 7040835
Flash Point 52°C (180°F)
Solubility Information Soluble in water.
Formula Weight 440.71
Percent Purity 97%
Refractive Index 1.3453
Sensitivity Moisture sensitive
Density 1.461 g/mL
Chemical Name or Material 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.